Anti-Monoamine Oxidase B/MAOB Rabbit Monoclonal Antibody [clone: EPR24131-79]

Supplier: Abcam
AB259928-100UL AB259928-20UL AB259928-1ML
ABCA_AB259928-100UEA 1376.18 CAD
ABCA_AB259928-100U ABCA_AB259928-20UL ABCA_AB259928-1ML
Anti-Monoamine Oxidase B/MAOB Rabbit Monoclonal Antibody [clone: EPR24131-79]
Antibodies
Rabbit monoclonal [EPR24131-79] to Monoamine Oxidase B/MAOB.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Monoamine Oxidase B/MAOB
Clonality: Monoclonal
Clone: EPR24131-79
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR