Anti-Cytokeratin 13 Rabbit Monoclonal Antibody [clone: EPR3672]

Supplier: Abcam
AB133340-1ML AB133340-100UL
ABCA_AB133340-1MLEA 9633.26 CAD
ABCA_AB133340-1ML ABCA_AB133340-100U
Anti-Cytokeratin 13 Rabbit Monoclonal Antibody [clone: EPR3672]
Antibodies
Rabbit monoclonal [EPR3672] to Cytokeratin 13.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 13
Clonality: Monoclonal
Clone: EPR3672
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR