Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y]

Supplier: Abcam
AB76020-1ML AB76020-20UL AB76020-250UL AB76020-100UL
ABCA_AB76020-1MLEA 7128.96 CAD
ABCA_AB76020-1ML ABCA_AB76020-20UL ABCA_AB76020-250UL ABCA_AB76020-100UL
Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y]
Antibodies
Rabbit monoclonal [EP1845Y] to Sodium Potassium ATPase - Plasma Membrane Loading Control.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human;Chinese hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR