Anti-VIM Rabbit Monoclonal Antibody [clone: EPR3776] (APC)

Supplier: Abcam
AB314250-10UL AB314250-100UL
ABCA_AB314250-10ULEA 267.02 CAD
ABCA_AB314250-10UL ABCA_AB314250-100U
Anti-VIM Rabbit Monoclonal Antibody [clone: EPR3776] (APC)
Antibodies
APC Rabbit monoclonal [EPR3776] to Vimentin - Cytoskeleton Marker.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: EPR3776
Conjugation: APC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR