Anti-p53 Rabbit Monoclonal Antibody [clone: EPR20416-124]

Supplier: Abcam
AB246223-100UL AB246223-1ML AB246223-20UL
ABCA_AB246223-100UEA 791.58 CAD
ABCA_AB246223-100U ABCA_AB246223-1ML ABCA_AB246223-20UL
Anti-p53 Rabbit Monoclonal Antibody [clone: EPR20416-124]
Antibodies
Rabbit monoclonal [EPR20416-124] to p53.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: EPR20416-124
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR