Anti-IL-3 Rabbit Antibody [clone: EPR20506-246]

Supplier: Abcam
AB244657-1MG AB244657-100UG
ABCA_AB244657-1MGEA 16109.68 CAD
ABCA_AB244657-1MG ABCA_AB244657-100U
Anti-IL-3 Rabbit Antibody [clone: EPR20506-246]
Antibodies
Rabbit monoclonal [EPR20506-246] to IL-3 - BSA and Azide free (Capture).

Type: Primary
Antigen: IL-3
Clonality:
Clone: EPR20506-246
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR