Anti-SIRP alpha Mouse Monoclonal Antibody [clone: OX-41]

Supplier: Abcam
AB307564-100UG AB307564-1MG
ABCA_AB307564-100UEA 1791.72 CAD
ABCA_AB307564-100U ABCA_AB307564-1MG
Anti-SIRP alpha Mouse Monoclonal Antibody [clone: OX-41]
Antibodies
Mouse monoclonal [OX-41] to SIRP alpha - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SIRP alpha
Clonality: Monoclonal
Clone: OX-41
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR