Anti-Influenza A virus PB1 protein Rabbit Monoclonal Antibody [clone: EPR28248-26]

Supplier: Abcam
AB313887-1ML AB313887-20UL AB313887-100UL
ABCA_AB313887-1MLEA 11287.52 CAD
ABCA_AB313887-1ML ABCA_AB313887-20UL ABCA_AB313887-100U
Anti-Influenza A virus PB1 protein Rabbit Monoclonal Antibody [clone: EPR28248-26]
Antibodies
Rabbit monoclonal [EPR28248-26] to Influenza A virus PB1 protein.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Influenza A virus PB1 protein
Clonality: Monoclonal
Clone: EPR28248-26
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Influenza A
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR