Anti-VISTA Rabbit Monoclonal Antibody [clone: EPR25068-124]

Supplier: Abcam
AB300042-100UL AB300042-1ML AB300042-20UL
ABCA_AB300042-100UEA 1297.18 CAD
ABCA_AB300042-100U ABCA_AB300042-1ML ABCA_AB300042-20UL
Anti-VISTA Rabbit Monoclonal Antibody [clone: EPR25068-124]
Antibodies
Rabbit monoclonal [EPR25068-124] to VISTA.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: VISTA
Clonality: Monoclonal
Clone: EPR25068-124
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR