Anti-CPT2 Rabbit Monoclonal Antibody [clone: EPR13626]

Supplier: Abcam
AB181114-1ML AB181114-100UL
ABCA_AB181114-1MLEA 11108.98 CAD
ABCA_AB181114-1ML ABCA_AB181114-100U
Anti-CPT2 Rabbit Monoclonal Antibody [clone: EPR13626]
Antibodies
Rabbit monoclonal [EPR13626] to CPT2 - C-terminal.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CPT2
Clonality: Monoclonal
Clone: EPR13626
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR