Anti-Cofilin-1 Rabbit Monoclonal Antibody [clone: EPR24514-41]

Supplier: Abcam
AB300184-1MG AB300184-100UG
ABCA_AB300184-1MGEA 15822.12 CAD
ABCA_AB300184-1MG ABCA_AB300184-100U
Anti-Cofilin-1 Rabbit Monoclonal Antibody [clone: EPR24514-41]
Antibodies
Rabbit monoclonal [EPR24514-41] to Cofilin-1 - BSA and Azide free (Capture).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning theMore Product Information
New Product
Order Now

SPECIFICATIONS

Antigen symbol Cofilin-1
Antigen name CFL
Environmentally Preferable
Antigen synonyms CFL1|18 kDa phosphoprotein|p18
Antibody type Primary
Clonality Monoclonal
Clone EPR24514-41
Reactivity
Host Rabbit
Isotype IgG
ELISA Yes
Immunogen The exact immunogen used to generate this antibody is proprietary information
Form Liquid
Molecular weight 18502 Da
Storage temperature Store at 4 °C
Shipping temperature 4 °C
Tested applications sELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR