Anti-delta 1 Catenin/CAS (phospho Y228) Rabbit Monoclonal Antibody [clone: E205]

Supplier: Abcam
AB32403-10UL AB32403-1ML AB32403-100UL
ABCA_AB32403-10ULEA 494.54 CAD
ABCA_AB32403-10UL ABCA_AB32403-1ML ABCA_AB32403-100UL
Anti-delta 1 Catenin/CAS (phospho Y228) Rabbit Monoclonal Antibody [clone: E205]
Antibodies
Rabbit monoclonal [E205] to delta 1 Catenin/CAS (phospho Y228).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BMX
Clonality: Monoclonal
Clone: Y396
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR