Anti-Histone H3.3 (tri methyl K79) Rabbit Monoclonal Antibody [clone: RM157]

Supplier: Abcam
AB275929-10UG AB275929-100UG
ABCA_AB275929-10UGEA 385.52 CAD
ABCA_AB275929-10UG ABCA_AB275929-100U
Anti-Histone H3.3 (tri methyl K79) Rabbit Monoclonal Antibody [clone: RM157]
Antibodies
Rabbit monoclonal [RM157] to Histone H3.3 (tri methyl K79).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3.3
Clonality: Monoclonal
Clone: RM157
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR