Anti-beta 3 Adrenergic Receptor Rabbit Monoclonal Antibody [clone: EPR25440-151]

Supplier: Abcam
AB300483-20UL AB300483-1ML AB300483-100UL
ABCA_AB300483-20ULEA 455.04 CAD
ABCA_AB300483-20UL ABCA_AB300483-1ML ABCA_AB300483-100U
Anti-beta 3 Adrenergic Receptor Rabbit Monoclonal Antibody [clone: EPR25440-151]
Antibodies
Rabbit monoclonal [EPR25440-151] to beta 3 Adrenergic Receptor.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: beta 3 Adrenergic Receptor
Clonality: Monoclonal
Clone: EPR25440-151
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR