Anti-IgG Goat Polyclonal Antibody (Gold, 40nm)

Supplier: Abcam
AB270694-1ML AB270694-200UL
ABCA_AB270694-1MLEA 802.64 CAD
ABCA_AB270694-1ML ABCA_AB270694-200U
Anti-IgG Goat Polyclonal Antibody (Gold, 40nm)
Antibodies
Goat Anti-Rabbit IgG (40nm Gold, 10 OD).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, 40nm
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR