Anti-beta 1 Spectrin Rabbit Monoclonal Antibody [clone: EPR6259]

Supplier: Abcam
AB129065-1ML AB129065-100UL
ABCA_AB129065-1MLEA 7892.1 CAD
ABCA_AB129065-1ML ABCA_AB129065-100U
Anti-beta 1 Spectrin Rabbit Monoclonal Antibody [clone: EPR6259]
Antibodies
Rabbit monoclonal [EPR6259] to beta 1 Spectrin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ALDH3A1
Clonality: Monoclonal
Clone: EPR7405
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR