Anti-IKK beta Rabbit Monoclonal Antibody [clone: Y466]

Supplier: Abcam
AB32135-1ML AB32135-100UL
ABCA_AB32135-1MLEA 11475.54 CAD
ABCA_AB32135-1ML ABCA_AB32135-100UL
Anti-IKK beta Rabbit Monoclonal Antibody [clone: Y466]
Antibodies
Rabbit monoclonal [Y466] to IKK beta.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IKK beta
Clonality: Monoclonal
Clone: Y466
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR