Anti-Glutamate receptor 4 Rabbit Monoclonal Antibody [clone: EPR24283-69]

Supplier: Abcam
AB280199-100UL AB280199-1ML AB280199-20UL
ABCA_AB280199-100UEA 1376.18 CAD
ABCA_AB280199-100U ABCA_AB280199-1ML ABCA_AB280199-20UL
Anti-Glutamate receptor 4 Rabbit Monoclonal Antibody [clone: EPR24283-69]
Antibodies
Rabbit monoclonal [EPR24283-69] to Glutamate receptor 4.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIF-1 alpha
Clonality: Monoclonal
Clone: 54/HIF-1a
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR