Anti-PDGFR alpha Rabbit Monoclonal Antibody [clone: EPR5480]

Supplier: Abcam
AB134123-1ML AB134123-20UL AB134123-100UL
ABCA_AB134123-1MLEA 11475.54 CAD
ABCA_AB134123-1ML ABCA_AB134123-20UL ABCA_AB134123-100U
Anti-PDGFR alpha Rabbit Monoclonal Antibody [clone: EPR5480]
Antibodies
Rabbit monoclonal [EPR5480] to PDGFR alpha.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PDGFR alpha
Clonality: Monoclonal
Clone: EPR5480
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR