Anti-DPP10 Rabbit Monoclonal Antibody

Supplier: Abcam
AB316772-10UL AB316772-20UL AB316772-100UL AB316772-1ML
ABCA_AB316772-10ULEA 415.54 CAD
ABCA_AB316772-10UL ABCA_AB316772-20UL ABCA_AB316772-100U ABCA_AB316772-1ML
Anti-DPP10 Rabbit Monoclonal Antibody
Antibodies
Rabbit monoclonal [EPR29237-246] to DPP10.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: DPP10
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR