Anti-STAT1 Rabbit Monoclonal Antibody [clone: EPR23049-111]

Supplier: Abcam
AB239360-20UL AB239360-100UL AB239360-1ML
ABCA_AB239360-20ULEA 425.02 CAD
ABCA_AB239360-20UL ABCA_AB239360-100U ABCA_AB239360-1ML
Anti-STAT1 Rabbit Monoclonal Antibody [clone: EPR23049-111]
Antibodies
Rabbit monoclonal [EPR23049-111] to STAT1 - ChIP Grade.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: EPR23049-111
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR