Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: EP1532Y]

Supplier: Abcam
AB317786-1ML AB317786-20UL AB317786-100UL
ABCA_AB317786-1MLEA 10099.36 CAD
ABCA_AB317786-1ML ABCA_AB317786-20UL ABCA_AB317786-100U
Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: EP1532Y]
Antibodies
Mouse monoclonal [EP1532Y] to Tyrosine Hydroxylase - Chimeric.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: EP1532Y
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR