Anti-LRRK2 (phospho S910) Rabbit Monoclonal Antibody [clone: UDD1 15(3)]

Supplier: Abcam
AB133449-100UL AB133449-1ML
ABCA_AB133449-100UEA 396.58 CAD
ABCA_AB133449-100U ABCA_AB133449-1ML
Anti-LRRK2 (phospho S910) Rabbit Monoclonal Antibody [clone: UDD1 15(3)]
Antibodies
Rabbit monoclonal [UDD1 15(3)] to LRRK2 (phospho S910).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LRRK2
Clonality: Monoclonal
Clone: UDD1 15(3)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR