Anti-Glutamate Receptor 1 (AMPA subtype) (phospho S831) Rabbit Monoclonal Antibody [clone: EPR1887]

Supplier: Abcam
AB109464-1ML AB109464-100UL
ABCA_AB109464-1MLEA 8723.18 CAD
ABCA_AB109464-1ML ABCA_AB109464-100U
Anti-Glutamate Receptor 1 (AMPA subtype) (phospho S831) Rabbit Monoclonal Antibody [clone: EPR1887]
Antibodies
Rabbit monoclonal [EPR1887] to Glutamate Receptor 1 (AMPA subtype) (phospho S831).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glutamate Receptor 1 (AMPA subtype)
Clonality: Monoclonal
Clone: EPR1887
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR