Anti-CD40 Rabbit Monoclonal Antibody [clone: EPR18005-35] (FITC (Fluorescein Isothiocyanate))

Supplier: Abcam
AB314951-10UL AB314951-100UL
ABCA_AB314951-10ULEA 366.56 CAD
ABCA_AB314951-10UL ABCA_AB314951-100U
Anti-CD40 Rabbit Monoclonal Antibody [clone: EPR18005-35] (FITC (Fluorescein Isothiocyanate))
Antibodies
FITC Rabbit monoclonal [EPR18005-35] to CD40.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: EPR18005-35
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR