Anti-RNF21/IFP1 Rabbit Monoclonal Antibody [clone: EPR11305(2)]

Supplier: Abcam
AB180130-100UL AB180130-1ML
ABCA_AB180130-100UEA 979.6 CAD
ABCA_AB180130-100U ABCA_AB180130-1ML
Anti-RNF21/IFP1 Rabbit Monoclonal Antibody [clone: EPR11305(2)]
Antibodies
Rabbit monoclonal [EPR11305(2)] to RNF21/IFP1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RNF21/IFP1
Clonality: Monoclonal
Clone: EPR11305(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR