Anti-Tau Rabbit Monoclonal Antibody [clone: EPR20508-35]

Supplier: Abcam
AB284978-100UG AB284978-1MG
ABCA_AB284978-100UEA 1624.24 CAD
ABCA_AB284978-100U ABCA_AB284978-1MG
Anti-Tau Rabbit Monoclonal Antibody [clone: EPR20508-35]
Antibodies
Rabbit monoclonal [EPR20508-35] to Tau - BSA and Azide free (Capture).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tau
Clonality: Monoclonal
Clone: EPR20508-35
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR