Anti-CK1 epsilon Mouse Monoclonal Antibody [clone: 1-Casein Kinase Iε]

Supplier: Abcam
AB302638-20UG AB302638-10UG AB302638-100UG AB302638-1MG
ABCA_AB302638-20UGEA 445.56 CAD
ABCA_AB302638-20UG ABCA_AB302638-10UG ABCA_AB302638-100U ABCA_AB302638-1MG
Anti-CK1 epsilon Mouse Monoclonal Antibody [clone: 1-Casein Kinase Iε]
Antibodies
Mouse monoclonal [1/Casein Kinase Ie] to CK1 epsilon.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CK1 epsilon
Clonality: Monoclonal
Clone: 1-Casein Kinase Iε
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR