Anti-LGALS3BP Rabbit Monoclonal Antibody [clone: EPR21757-33]

Supplier: Abcam
AB217572-100UL AB217572-1ML
ABCA_AB217572-100UEA 1000.14 CAD
ABCA_AB217572-100U ABCA_AB217572-1ML
Anti-LGALS3BP Rabbit Monoclonal Antibody [clone: EPR21757-33]
Antibodies
Rabbit monoclonal [EPR21757-33] to LGALS3BP.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LGALS3BP
Clonality: Monoclonal
Clone: EPR21757-33
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR