Anti-ZAP70 Mouse Monoclonal Antibody [clone: 29-ZAP70 Kinase]

Supplier: Abcam
AB302821-100UG AB302821-20UG AB302821-1MG
ABCA_AB302821-100UEA 1287.7 CAD
ABCA_AB302821-100U ABCA_AB302821-20UG ABCA_AB302821-1MG
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 29-ZAP70 Kinase]
Antibodies
Mouse monoclonal [29/ZAP70 Kinase] to ZAP70.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: 29-ZAP70 Kinase
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR