Anti-Syntaxin 1a Rabbit Monoclonal Antibody [clone: EPR11072(B)]

Supplier: Abcam
AB170890-1ML AB170890-100UL
ABCA_AB170890-1MLEA 9544.78 CAD
ABCA_AB170890-1ML ABCA_AB170890-100U
Anti-Syntaxin 1a Rabbit Monoclonal Antibody [clone: EPR11072(B)]
Antibodies
Rabbit monoclonal [EPR11072(B)] to Syntaxin 1a.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Syntaxin 1a
Clonality: Monoclonal
Clone: EPR11072(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR