Anti-CD11c Mouse Monoclonal Antibody [clone: KB90] (PE (Phycpoerythrin))

Supplier: Abcam
AB316176-100UL AB316176-10UL
ABCA_AB316176-100UEA 1248.2 CAD
ABCA_AB316176-100U ABCA_AB316176-10UL
Anti-CD11c Mouse Monoclonal Antibody [clone: KB90] (PE (Phycpoerythrin))
Antibodies
PE Mouse monoclonal [KB90] to CD11c.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: KB90
Conjugation: PE (Phycpoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR