Anti-Uroplakin III Rabbit Polyclonal Antibody

Supplier: Abcam
AB82173-1ML AB82173-200UL
ABCA_AB82173-1MLEA 1663.74 CAD
ABCA_AB82173-1ML ABCA_AB82173-200UL
Anti-Uroplakin III Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Uroplakin III.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Uroplakin III
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR