Anti-Glycerol kinase Rabbit Monoclonal Antibody [clone: EPR6567]

Supplier: Abcam
AB126599-10UL AB126599-100UL AB126599-1ML
ABCA_AB126599-10ULEA 485.06 CAD
ABCA_AB126599-10UL ABCA_AB126599-100U ABCA_AB126599-1ML
Anti-Glycerol kinase Rabbit Monoclonal Antibody [clone: EPR6567]
Antibodies
Rabbit monoclonal [EPR6567] to Glycerol kinase.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glycerol kinase
Clonality: Monoclonal
Clone: EPR6567
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR