Anti-Diphtheria Toxin Human Monoclonal Antibody [clone: IGX3492]

Supplier: Abcam
AB209329-1ML AB209329-100UL
ABCA_AB209329-1MLEA 8901.72 CAD
ABCA_AB209329-1ML ABCA_AB209329-100U
Anti-Diphtheria Toxin Human Monoclonal Antibody [clone: IGX3492]
Antibodies
Human monoclonal [IGX3492] to Diphtheria Toxin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Diphtheria Toxin
Clonality: Monoclonal
Clone: IGX3492
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR