Anti-HSC70 Interacting Protein HIP Rabbit Monoclonal Antibody [clone: EP3264]

Supplier: Abcam
AB284733-1MG AB284733-100UG
ABCA_AB284733-1MGEA 17792.38 CAD
ABCA_AB284733-1MG ABCA_AB284733-100U
Anti-HSC70 Interacting Protein HIP Rabbit Monoclonal Antibody [clone: EP3264]
Antibodies
Rabbit monoclonal [EP3264] to HSC70 Interacting Protein HIP - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: HSC70 Interacting Protein HIP
Clonality: Monoclonal
Clone: EP3264
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR