Anti-Polyethylene glycol Rabbit Monoclonal Antibody [clone: PEG-B-47b]

Supplier: Abcam
AB271853-100UG AB271853-1MG
ABCA_AB271853-100UEA 1791.72 CAD
ABCA_AB271853-100U ABCA_AB271853-1MG
Anti-Polyethylene glycol Rabbit Monoclonal Antibody [clone: PEG-B-47b]
Antibodies
Rabbit monoclonal [PEG-B-47b] to Polyethylene glycol - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Polyethylene glycol
Clonality: Monoclonal
Clone: PEG-B-47b
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR