Anti-Carbonic Anhydrase 7/CA7 Rabbit Monoclonal Antibody [clone: EPR13304]

Supplier: Abcam
AB191408-1ML AB191408-100UL
ABCA_AB191408-1MLEA 8623.64 CAD
ABCA_AB191408-1ML ABCA_AB191408-100U
Anti-Carbonic Anhydrase 7/CA7 Rabbit Monoclonal Antibody [clone: EPR13304]
Antibodies
Rabbit monoclonal [EPR13304] to Carbonic Anhydrase 7/CA7.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Carbonic Anhydrase 7/CA7
Clonality: Monoclonal
Clone: EPR13304
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR