Anti-SAP97 Rabbit Monoclonal Antibody [clone: EPR26467-123]

Supplier: Abcam
AB300481-10UL AB300481-1ML AB300481-20UL AB300481-100UL
ABCA_AB300481-10ULEA 415.54 CAD
ABCA_AB300481-10UL ABCA_AB300481-1ML ABCA_AB300481-20UL ABCA_AB300481-100U
Anti-SAP97 Rabbit Monoclonal Antibody [clone: EPR26467-123]
Antibodies
Rabbit monoclonal [EPR26467-123] to SAP97.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SAP97
Clonality: Monoclonal
Clone: EPR26467-123
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR