Anti-Prolactin Receptor/PRL-R Rabbit Monoclonal Antibody [clone: 204] (APC (Allophycocyanin))

Supplier: Abcam
AB275715-100TEST AB275715-25TEST
ABCA_AB275715-100TEA 1099.68 CAD
ABCA_AB275715-100T ABCA_AB275715-25TE
Anti-Prolactin Receptor/PRL-R Rabbit Monoclonal Antibody [clone: 204] (APC (Allophycocyanin))
Antibodies
APC Rabbit monoclonal [204] to Prolactin Receptor/PRL-R.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Prolactin Receptor/PRL-R
Clonality: Monoclonal
Clone: 204
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR