Anti-SIRP alpha Mouse Monoclonal Antibody [clone: 3] (FITC (Fluorescein Isothiocyanate))

Supplier: Abcam
AB289909-25TEST AB289909-100TEST
ABCA_AB289909-25TEEA 376.04 CAD
ABCA_AB289909-25TE ABCA_AB289909-100T
Anti-SIRP alpha Mouse Monoclonal Antibody [clone: 3] (FITC (Fluorescein Isothiocyanate))
Antibodies
FITC Mouse monoclonal [03] to SIRP alpha.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SIRP alpha
Clonality: Monoclonal
Clone: 3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR