Anti-CD18 Rat Monoclonal Antibody [clone: C71/16]

Supplier: Abcam
AB19580-10UG AB19580-100UG AB19580-1MG
ABCA_AB19580-10UGEA 346.02 CAD
ABCA_AB19580-10UG ABCA_AB19580-100UG ABCA_AB19580-1MG
Anti-CD18 Rat Monoclonal Antibody [clone: C71/16]
Antibodies
Rat monoclonal [C71/16] to CD18.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: C71/16
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR