Anti-LRRK2 Rabbit Monoclonal Antibody [clone: UDD3 30(12)]

Supplier: Abcam
AB133518-1ML AB133518-100UL
ABCA_AB133518-1MLEA 5326.18 CAD
ABCA_AB133518-1ML ABCA_AB133518-100U
Anti-LRRK2 Rabbit Monoclonal Antibody [clone: UDD3 30(12)]
Antibodies
Rabbit monoclonal [UDD3 30(12)] to LRRK2.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LRRK2
Clonality: Monoclonal
Clone: UDD3 30(12)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR