Anti-TRIM46 Rabbit Monoclonal Antibody [clone: EPR26957-34]

Supplier: Abcam
AB307967-1ML AB307967-20UL AB307967-100UL
ABCA_AB307967-1MLEA 11287.52 CAD
ABCA_AB307967-1ML ABCA_AB307967-20UL ABCA_AB307967-100U
Anti-TRIM46 Rabbit Monoclonal Antibody [clone: EPR26957-34]
Antibodies
Rabbit monoclonal [EPR26957-34] to TRIM46.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TRIM46
Clonality: Monoclonal
Clone: EPR26957-34
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR