Anti-IL-8 Mouse Monoclonal Antibody [clone: O-IL8-15]

Supplier: Abcam
AB282027-100UG AB282027-1MG
ABCA_AB282027-100UEA 1791.72 CAD
ABCA_AB282027-100U ABCA_AB282027-1MG
Anti-IL-8 Mouse Monoclonal Antibody [clone: O-IL8-15]
Antibodies
Mouse monoclonal [O-IL8-15] to IL-8 - Chimeric – BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type:
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pigeon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR