Anti-TARC/CCL17 Rabbit Monoclonal Antibody [clone: YCA-R21766-165 H1L2]

Supplier: Abcam
AB288617-1MG AB288617-100UG
ABCA_AB288617-1MGEA 15436.6 CAD
ABCA_AB288617-1MG ABCA_AB288617-100U
Anti-TARC/CCL17 Rabbit Monoclonal Antibody [clone: YCA-R21766-165 H1L2]
Antibodies
Rabbit monoclonal [YCA-R21766-165 H1L2] to TARC/CCL17 - BSA and Azide free (Detector).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TARC/CCL17
Clonality: Monoclonal
Clone: YCA-R21766-165 H1L2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR