Anti-TCP1 delta Rabbit Monoclonal Antibody [clone: EPR8495(B)]

Supplier: Abcam
AB129072-100UL AB129072-1ML
ABCA_AB129072-100UEA 891.12 CAD
ABCA_AB129072-100U ABCA_AB129072-1ML
Anti-TCP1 delta Rabbit Monoclonal Antibody [clone: EPR8495(B)]
Antibodies
Rabbit monoclonal [EPR8495(B)] to TCP1 delta.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TCP1 delta
Clonality: Monoclonal
Clone: EPR8495(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR