Anti-Tubulin (acetyl K40) Rabbit Monoclonal Antibody [clone: EPR13797]

Supplier: Abcam
AB250822-1MG AB250822-100UG
ABCA_AB250822-1MGEA 16158.66 CAD
ABCA_AB250822-1MG ABCA_AB250822-100U
Anti-Tubulin (acetyl K40) Rabbit Monoclonal Antibody [clone: EPR13797]
Antibodies
Rabbit monoclonal [EPR13797] to Tubulin - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tubulin
Clonality: Monoclonal
Clone: EPR13797
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR