Anti-Tyrosine Hydroxylase Rabbit Recombinant Multiclonal Antibody [clone: RM1085]

Supplier: Abcam
AB315252-20UL AB315252-1ML AB315252-100UL
ABCA_AB315252-20ULEA 366.56 CAD
ABCA_AB315252-20UL ABCA_AB315252-1ML ABCA_AB315252-100U
Anti-Tyrosine Hydroxylase Rabbit Recombinant Multiclonal Antibody [clone: RM1085]
Antibodies
Rabbit recombinant multiclonal [RM1085] to Tyrosine Hydroxylase.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Recombinant Multiclonal
Clone: RM1085
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR