Anti-Histone H2A.X Rabbit Monoclonal Antibody [clone: EPR22820-23]

Supplier: Abcam
AB229914-1ML AB229914-100UL AB229914-20UL
ABCA_AB229914-1MLEA 7435.48 CAD
ABCA_AB229914-1ML ABCA_AB229914-100U ABCA_AB229914-20UL
Anti-Histone H2A.X Rabbit Monoclonal Antibody [clone: EPR22820-23]
Antibodies
Rabbit monoclonal [EPR22820-23] to Histone H2A.X - ChIP Grade.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H2A.X
Clonality: Monoclonal
Clone: EPR22820-23
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR